missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Epi5™ Episomal iPSC Reprogramming Kit

Catalog No. a15960
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15960 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A15960 Supplier Invitrogen™ Supplier No. A15960
Only null left

Epi5™ Episomal iPSC Reprogramming Kit

The Epi5™ Episomal iPSC Reprogramming Kit provides an easy-to-use, highly efficient set of 5 episomal vectors designed by Dr. Okita in the laboratory of Professor Yamanaka at the Center for iPS Cell Research and Application (CiRA), Kyoto University. This system produces transgene-free, virus-free human induced pluripotent stem cells (iPSCs) with efficiencies in the range of 0.04% to 0.3%, depending upon the cell type being reprogrammed.

Features of the Epi5™ Episomal iPSC Reprogramming Kit include:

  • Easy to use and highly efficient—no need for small molecules during reprogramming and the added p53 suppression provides enhanced iPSC generation (1)
  • Usable for all stages of your iPSC research—transgene-free and viral-free reprogramming allows use from basic to pre-clinical research
  • Flexibility in media systems—can be used in either feeder-free or feeder-based media systems

    Create Transgene and Virus-free iPSCs
    Episomal vectors are a well-described system for producing transgene-free, virus-free iPSCs, providing a source of iPSCs for all stages of your pluripotent stem cell research. Other reprogramming methods, such as lentivirus, contain transgenes that can integrate into the host genome, potentially disrupting the genome or causing unpredictable results.

    As oriP/EBNA1 vectors, these episomal vectors contain 5 reprogramming factors (Oct4, Sox2, Lin28, Klf4, and L-Myc) and replicate extra-chromosomally only once per cell cycle. At this replication rate, the episomes are lost at a rate of approximately 5% per cell generation. This system shows enhanced iPSC generation through p53 suppression, and the inclusion of L-Myc has been shown to be more potent and specific then c-Myc during human iPSC generation (1).

    Easy-to-Use System
    This 2-vial kit provides the optimal system for episomal vector reprogramming and eliminates the need for use of small molecules in the reprogramming medium. The episomal vectors are introduced into the cell by electroporation. We recommend use of the Neon™Transfection System for electroporation, allowing flexibility in the volume and thus number of cells needed for reprogramming.

    Flexibility in Selection of Media Systems
    The Epi5™ Episomal iPSC Reprogramming Kit can be used with multiple cell types and media systems. Protocols for fibroblasts and CD34+ blood cells are available in either feeder or feeder-free based reprogramming.

    1. Okita K, Matsumara Y, Sato Y, et al. A more efficient method to generate integration free human iPS cells. Nature Methods 8, 409-412, 2011.
  • Specifications

    Product Line Epi5
    Quantity 1 kit
    For Use With (Application) Stem Cell Engineering & Reprogramming
    For Use With (Equipment) Electroporator, Neon Transfection System
    Product Type Episomal iPSC Reprogramming Kit
    Promoter CAG
    Protein Tag None
    Replication Origin pUC Origin
    Vector EBNA OriP
    Content And Storage 2 tubes. Store at -5 to -20°C.
    Selection Agent (Eukaryotic) None
    Cloning Method None
    Reporter Gene None
    Key Functions el
    Delivery Type Transfection
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.