Recombinant Protein;1MG;Human Ephrin-A4, Fc Tag (EA4-H5250) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Gly 171 (Accession # AAI07484).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 43.0 kDa. The protein migrates as 34 kDa and 48 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human EphA4, His Tag (Cat. No. EP4-H52H8) at 5 g/mL (100 L/well) can bind Human Ephrin-A4, Fc Tag (Cat. No. EA4-H5250) with a linear range of 0.6-5 ng/mL (QC tested).