Ace Glass Arsenic Limit Test, European Pharmacopeia, Complete with125mL Erlenmeyer, chimney tube, bent tube, springs and3mL marked test tube
Catalog No. NC1474014
Arsenic limit test apparatus as described in the European Pharmacopeia 9.4, chapter 2.4.2. Complete apparatus consists of 125mL erlenmeyer flask, 24/40 chimney tube with flat ground o-ring joint, bent tube with flat ground o-ring joint and 1mm drip tip, test tube and springs for securing the chimney tube to the bent tube.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Arsenic limit test apparatus as described in the European Pharmacopeia 9.4, chapter 2.4.2. Complete apparatus consists of 125mL erlenmeyer flask, 24/40 chimney tube with flat ground o-ring joint, bent tube with flat ground o-ring joint and 1mm drip tip, test tube and springs for securing the chimney tube to the bent tube.