MW 647.9 g/mol. A red fluorescent dye from the xanthene family used as an acidic red stain for cell cytoplasm. Reduces the affinity of Mg+-ATPase for ATP and increases the enzyme affinity for Mg+ at concentrations of 10-50 M. At higher concentrations near 100 M the affinity for Mg+-ATPase for the ion-activator has been shown to be reduced.
The product is subject to the following: Abcam Restricted Use Statement