missing translation for 'onlineSavingsMsg'
Learn More

Eosin Y Stain, 0.25% (w/v) in 57% (v/v) Alcohol, Acidified Working Solution, Ricca Chemical

Catalog No. 284516
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
284516 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 284516 Supplier Ricca Chemical Company Supplier No. 284516

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Eosin Y Stain
Name Note Acidified Working Solution
CAS 108-10-1, 17372-87-1, 67-63-0, 67-56-1, 64-19-7
CAS Min % 66.0, 28.79, 1.86, 0.56, 0.5
CAS Max % 69.0, 29.97, 1.94, 0.59, 0.52
Concentration 0.25% (w/v)
Packaging Natural Poly Bottle
Solvent or Matrix 57% (v/v) Alcohol
Color Orange/Red
pH 4.65
Quantity 500 mL
Physical Form Liquid
Percent Purity 0.25% W/V
Grade Indicator
Show More Show Less