missing translation for 'onlineSavingsMsg'
Learn More

Sigma-Aldrich Eosin Y Solution, Alcoholic

Catalog No. 01288059
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01288059 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01288059 Supplier Sigma-Aldrich Supplier No. HT1101321L

General purpose cytoplasmic counterstain. Used with hematoxylin and eosin staining.

Certified Eosin Y, 0.5% (w/v) in acidified 90% ethanol.

  • Form: solution
  • Shelf life: expiry date on the label
  • IVD: for in vitro diagnostic use
  • Concentration: 0.5% (w/v) in acidified ethanol
  • Applications: hematology and histology
  • Storage temperature: room temperature

Specifications

Quantity 1 L
Product Type Eosin Y