missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Eosin Y, pure, Indicator grade

CAS: 17372-87-1 | C20H12Br4Na2O8 | 745.904 g/mol

Supplier:  Thermo Scientific Chemicals 201931000

Quantity 100 g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC201931000


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

17372-87-1
745.904
MASXMTNVNASWNH-UHFFFAOYSA-L
91886399
C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
C20H12Br4Na2O8
MFCD00005040
2';,4';,5';,7';-Tetrabromofluorescein, disodium salt, Eosin Yellowish, 2';, 4';, 5';
disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate

Specifications

Eosin Y
17372-87-1
MFCD00005040
15, 3658
Solubility in water: freely soluble. Other solubilities: sparingly soluble in alcohol, insoluble in diethyl ether
C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate
91886399
691.85
Pure
1100 min. (lambda max, 0.001M Na2CO3) on dried substance
Complies
Brown/Red
Fine Crystalline Powder
>300.0°C
C20H12Br4Na2O8
19, IV, 2917
2';,4';,5';,7';-Tetrabromofluorescein, disodium salt, Eosin Yellowish, 2';, 4';, 5';
MASXMTNVNASWNH-UHFFFAOYSA-L
Authentic
745.904
511 to 519nm
10% max.
Glass bottle
1.018
1.0180g/mL
100 g
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
Documents

Documents

Promotions

Promotions