missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Eosin Y, pure, high purity, biological stain

Catalog No. AC152880250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC152880250 25 g Glass bottle
AC152881000 100 g Glass bottle
AC152885000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC152880250 Supplier Thermo Scientific Chemicals Supplier No. 152880250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17372-87-1 | C20H12Br4Na2O8 | 745.904 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 17372-87-1
Molecular Formula C20H12Br4Na2O8
Molecular Weight (g/mol) 745.904
MDL Number MFCD00005040
InChI Key MASXMTNVNASWNH-UHFFFAOYSA-L
Synonym 2',4',5',7'-Tetrabromofluorescein, disodium salt, Eosin Yellowish, 2', 4', 5'
PubChem CID 91886399
IUPAC Name disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate
SMILES C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]

Specifications

Melting Point >300.0°C
Dye Content 90% min. (on dry substance)
Infrared Spectrum Authentic
Loss on Drying 10% max.
Packaging Glass bottle
Specific Gravity 1.018
Beilstein 19, IV, 2917
Density 1.0180g/mL
Merck Index 15, 3658
Color Brown to Red
Solubility Information Solubility in water: freely soluble. Other solubilities: sparingly soluble in alcohol, insoluble in diethyl ether
Quantity 25 g
Physical Form Fine Crystalline Powder
Formula Weight 691.85
Grade Pure
Chemical Name or Material Eosin Y, High purity biological stain
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only