missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Eosin Y, pure, certified

CAS: 17372-87-1 | C20H12Br4Na2O8 | 745.904 g/mol

Supplier:  Thermo Scientific Chemicals 409430250

Catalog No. AC409430250


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

Eosin Y
1.21 to 1.77
C20H12Br4Na2O8
19, IV, 2917
2',4',5',7'-Tetrabromofluorescein, disodium salt, Eosin Yellowish, C.I. 45380, 2', 4', 5'
Authentic
disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate
91886399
691.85
Glass bottle
Brown to Brown/Green
Powder
17372-87-1
80% min.
MFCD00005040
15, 3658
MASXMTNVNASWNH-UHFFFAOYSA-L
C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
745.904
515 to 518nm
Pure
1.0180g/mL
25 g

Chemical Identifiers

17372-87-1
745.904
MASXMTNVNASWNH-UHFFFAOYSA-L
91886399
C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
C20H12Br4Na2O8
MFCD00005040
2',4',5',7'-Tetrabromofluorescein, disodium salt, Eosin Yellowish, C.I. 45380, 2', 4', 5'
disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only