missing translation for 'onlineSavingsMsg'
Learn More

Eosin Y, Certified, Ricca Chemical

Supplier:  Ricca Chemical Company RDCE005025F1

Catalog No. RDCE005025F


Only null left

Description

Description

Specifications

Specifications

Eosin Y
98.0
C20H12Br4Na2O8
C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
745.904
Indicator
Brown/Red
17372-87-1
100.0
MASXMTNVNASWNH-UHFFFAOYSA-L
disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate
91886399
Pouch
25 g

Chemical Identifiers

17372-87-1
745.904
91886399
C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
C20H12Br4Na2O8
MASXMTNVNASWNH-UHFFFAOYSA-L
disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 60 months

SDS
Documents

Documents

Promotions

Promotions