missing translation for 'onlineSavingsMsg'
Learn More

Eosin Y (Certified Biological Stain), Fisher Chemical™

Catalog No. p-22190
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
E51125 25 g Glass Bottle
E511100 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E51125 Supplier Thermo Fisher Scientific Supplier No. E511 25
Only null left

Chemical Identifiers

CAS 17372-87-1
Molecular Formula C20H12Br4Na2O8
Molecular Weight (g/mol) 745.904
MDL Number MFCD00005040
InChI Key MASXMTNVNASWNH-UHFFFAOYSA-L
Synonym FD&C Red No. 22, Acid Red 87
PubChem CID 91886399
IUPAC Name disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate
SMILES C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]

Specifications

Bio. Stain Comm. Certification Pass Test
Dye Content 88% min.
Formula Weight 691.85
Grade Certified Biological Stain
Packaging Glass Bottle
Color Index No. 45380
Color Brown/Red
Quantity 25 g
Physical Form Solid
Chemical Name or Material Eosin Y