missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eosin Y, 1% Alcoholic, LabChem™
Specifications
Specifications
| Physical Form | Liquid |
| Chemical Name or Material | Eosin Y |
| Name Note | 1% Alcoholic |
| CAS Max % | 89.5, 5.25, 4.25, 1 |
| CAS | 67-63-0, 67-56-1, 17372-87-1 |
| Packaging | Poly Bottle |
| Solubility | Soluble in water |
| Decomposition Information | Carbon monoxide; Carbon dioxide; Hydrogen bromide |
| Color | Green |
| Quantity | 1 L |
| Show More |