missing translation for 'onlineSavingsMsg'
Learn More

Eosin Y, 1% Alcoholic, LabChem™

Catalog No. LC140301
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC140301 500 mL
LC140302 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC140301 Supplier LabChem Supplier No. LC140301
Only null left

Specifications

Physical Form Liquid
Chemical Name or Material Eosin Y
Name Note 1% Alcoholic
CAS Max % 89.5, 5.25, 4.25, 1
CAS 67-63-0, 67-56-1, 17372-87-1
Packaging Poly Bottle
Solubility Soluble in water
Decomposition Information Carbon monoxide; Carbon dioxide; Hydrogen bromide
Color Green
Quantity 500 mL
Linear Formula C20H6Br4O5·2Na
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
Formula Weight 691.86
Show More Show Less