missing translation for 'onlineSavingsMsg'
Learn More

Eosin B

Catalog No. AAA1737722
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1737722 100 g
AAA1737714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1737722 Supplier Thermo Scientific Chemicals Supplier No. A1737722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 548-24-3 | C20H6Br2N2Na2O9

Eosin B sodium salt is a cell and tissue stain. Eosin B sodium salt is used as a counter stain for collagen, as a tissue stain for cell granules, nuclei, and microorganisms when coupled with Azure A (sc-203729), and as a stain for cell differentiation of the anterior pituitary.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Eosin B sodium salt is a cell and tissue stain. Eosin B sodium salt is used as a counter stain for collagen, as a tissue stain for cell granules, nuclei, and microorganisms when coupled with Azure A (sc-203729), and as a stain for cell differentiation of the anterior pituitary.

Solubility
Soluble in water (390 g/L at 20°C), and ethanol.

Notes
Store at room temperature. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 548-24-3
Molecular Formula C20H6Br2N2Na2O9
MDL Number MFCD00005041
Synonym C.I. 45400; 4,5-Dibromo-2,7-dinitrofluorescein disodium salt

Specifications

Quantity 100 g
Solubility Information Soluble in water (390g/L at 20°C),and ethanol.
Formula Weight 624.08
Chemical Name or Material Eosin B
Hazard Category H318
Precautionary Statement P280-P305+P351+P338-P310
EINECSNumber 208-943-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only