missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Eosin B, pure, high purity, biological stain

Eosin B, C20H6Br2N2Na2O9, CAS Number-548-24-3, 4', 5'-Dibromo-2', 7'-dinitrofluorescein, 4',5'-Dibromo-2',7'-dinitrofluorescein, disodium salt, Eosin Bluish, 100g, 0.96 to 1.22, 19, 233, Brown, 208-943-1, 624.06, Pure, Danger, GHS H Statement: Causes serious eye damage. | CAS: 548-24-3 | C20H6Br2N2Na2O9 | 624.06 g/mol

Supplier:  Thermo Scientific Chemicals 229650250

 View more versions of this product

Catalog No. AC229650250

Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

Eosin B
0.96 to 1.22
MFCD00005041
15, 3657
Authentic
624.06
Glass bottle
25 g
548-24-3
C20H6Br2N2Na2O9
19, 233
4',5'-Dibromo-2',7'-dinitrofluorescein, disodium salt, 4', 5'-Dibromo-2', 7'-dinitrofluorescein, Eosin Bluish
511 to 520nm (in water + 1% Na2CO3)
Pure
Brown
Crystalline Powder

Chemical Identifiers

548-24-3
C20H6Br2N2Na2O9
MFCD00005041
4',5'-Dibromo-2',7'-dinitrofluorescein, disodium salt, 4', 5'-Dibromo-2', 7'-dinitrofluorescein, Eosin Bluish
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 208-943-1

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only