Learn More
AH EOMES APC-EFLUOR 780 25 TS
Mouse Monoclonal Antibody
Supplier: Thermo Fisher Scientific 47487741
Description
Description: This WD1928 antibody recognizes Eomesodermin (Eomes), also known as T-box brain 2 (TBR2). Eomes is a T-box transcription factor that is highly homologous to T-bet, which is essential during trophoblast development and gastrulation in most vertebrates. In the immune system, Eomes controls the differentiation of effector and memory CD8+ T cells, as well as natural killer (NK) cells. Expression of Eomes in these cells correlates with high expression of CD122, the common beta-chain of the IL-2R and IL-15R. Applications Reported: This WD1928 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This WD1928 antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of normal human peripheral blood cells using the Foxp3/Transcription Factor Staining Buffer Set (cat. 00-5523) and protocol. Refer to Best Protocols for Staining Protocol (refer to Protocol B: One-step protocol for intracellular (nuclear) proteins). This can be used at 5 μL (0.03 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. APC-eFluor™ 780 emits at 780 nm and is excited with the Red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.
EOMES Monoclonal antibody specifically detects EOMES in Human, Porcine samples. It is validated for Flow Cytometry.Chemical Identifiers
| EOMES | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
| C77258; EOMES; eomesodermin; eomesodermin homolog; eomesodermin homolog (Xenopus laevis); T-box brain protein 2; T-box transcription factor; TBR2; TBR-2; T-brain-2 | |
| Eomes | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Eomes |
| WD1928 | |
| Flow Cytometry | |
| APC-eFluor 780 | |
| Mouse | |
| Human, Porcine | |
| O95936 | |
| 100622125, 8320 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
Specifications
| EOMES | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
| O95936 | |
| Eomes | |
| Affinity chromatography | |
| RUO | |
| 100622125, 8320 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| WD1928 | |
| APC-eFluor 780 | |
| Eomes | |
| C77258; EOMES; eomesodermin; eomesodermin homolog; eomesodermin homolog (Xenopus laevis); T-box brain protein 2; T-box transcription factor; TBR2; TBR-2; T-brain-2 | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Human, Porcine | |
| Antibody | |
| IgG1 κ |
For Research Use Only.