missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Covidien EnviroStar™ Noninfectious Waste Sharps Disposal Containers
Manufactured with recyled materials, these containers are designed to reduce injuries and disposal costs for non-contaminated sharps
$88.43 - $88.43
Specifications
| Color | Green |
|---|---|
| Model | EnviroStar |
| Autoclavable | Autoclavable |
| For Use With (Application) | For sharps not infected with blood-borne pathogens |
| Catalog Number | Mfr. No. | Capacity (English) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (English) | Price | Quantity | |||||
|
14-830-108
|
Covidien
8781 |
8 gal. |
Each for $88.43
Case of 10 Each for $884.30
|
|
|||||
Description
- Designed to contain sharps that can cause a percutaneous tear but are not infected with bloodborne pathogens, such as light bulbs, broken slides or Petri dishes, and glass lab bottles
- Environmental green container color helps to differentiate from other disposal containers and promotes proper waste segregation
- Made with environmentally friendly recycled material, enhancing Reduce-Recycle-Reuse programs
Specifications
| Green | |
| EnviroStar | |
| Autoclavable | |
| For sharps not infected with blood-borne pathogens |