Learn More
Environmental Express™ StepSaver™ Device for Solid-Phase Extraction: Tubes
Catalog No. 09735123
Change view
Click to view available options
For Use With (Equipment):
StepSaver
Catalog No. For Use With (Equipment)
09735123 StepSaver
1 options
Catalog No. 09735123 Supplier Environmental Express™ Supplier No. G1025
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Requires
Manifold, SPE extraction disk and Na2SO4 drying cartridge must be purchased separately.
Simplifies U.S. EPA Method 1664
  • Positions the collection vessel outside of the filter flask or manifold and utilizes a vacuum bridge to connect the vacuum source
  • Luer adapter, located in the solvent collection arm, permits the use of an inline Na2SO4 drying cartridge
  • Allows the collection of the solvent with eluted oil or grease directly into a preweighed collection flask, thus eliminating the disassembly or assembly of glassware
  • Already dried with the Na2SO4, the eluent can be directly evaporated to obtain the weight of the extracted oil or grease
  • Two-way, 120° PTFE-resin stopcock prevents accidental sample loss; attaches to a filter manifold or filtration flask through the standard taper 40/35 ground glass joint on the bottom of the apparatus
  • Designed for use with 47mm solid-phase extraction disks
  • Manufactured of borosilicate glass with a stainless-steel filter screen
Specifications
For Use With (Equipment) StepSaver
Type Stopcock
Includes Washer, O-ring and nut
Material PTFE