missing translation for 'onlineSavingsMsg'
Learn More

Entecavir Monohydrate 98.0+%, TCI America™

Catalog No. E089950MG
Change view
Click to view available options
Quantity:
50 mg
200 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E089950MG 50 mg
E0899200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E089950MG Supplier TCI America Supplier No. E089950MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 209216-23-9
Molecular Formula C12H17N5O4
Molecular Weight (g/mol) 295.299
MDL Number MFCD09754448
InChI Key YXPVEXCTPGULBZ-WQYNNSOESA-N
Synonym entecavir hydrate, entecavir monohydrate, baraclude tn, entecavir usan, entecavir hydrate jan, baraclude, 9-1s,3r,4s-4-hydroxy-3-hydroxymethyl-2-methylenecyclopentyl guanine monohydrate, entecavir inn, 2-amino-9-1s,3r,4s-4-hydroxy-3-hydroxymethyl-2-methylidenecyclopentyl-1,9-dihydro-6h-purin-6-one-water 1/1
PubChem CID 16052026
ChEBI CHEBI:59902
IUPAC Name 2-amino-9-[(1S,3R,4S)-4-hydroxy-3-(hydroxymethyl)-2-methylidenecyclopentyl]-3H-purin-6-one;hydrate
SMILES C=C1C(CC(C1CO)O)N2C=NC3=C2NC(=NC3=O)N.O

Specifications

Melting Point 259°C
Color White
Quantity 50 mg
Formula Weight 277.28
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Entecavir Monohydrate