missing translation for 'onlineSavingsMsg'
Learn More

Entecavir monohydrate, 99%, Thermo Scientific Chemicals

Catalog No. AC461050010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461050010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC461050010 Supplier Thermo Scientific Chemicals Supplier No. 461050010

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 209216-23-9
Molecular Formula C12H15N5O3·H20
Molecular Weight (g/mol) 295.29
InChI Key YXPVEXCTPGULBZ-WQYNNSOESA-N
Synonym entecavir hydrate, entecavir monohydrate, baraclude tn, entecavir usan, entecavir hydrate jan, baraclude, 9-1s,3r,4s-4-hydroxy-3-hydroxymethyl-2-methylenecyclopentyl guanine monohydrate, entecavir inn, 2-amino-9-1s,3r,4s-4-hydroxy-3-hydroxymethyl-2-methylidenecyclopentyl-1,9-dihydro-6h-purin-6-one-water 1/1
PubChem CID 16052026
ChEBI CHEBI:59902
IUPAC Name 2-amino-9-[(1S,3R,4S)-4-hydroxy-3-(hydroxymethyl)-2-methylidenecyclopentyl]-3H-purin-6-one;hydrate
SMILES C=C1C(CC(C1CO)O)N2C=NC3=C2NC(=NC3=O)N.O

Specifications

Melting Point 259.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: 0,24 g/100mL. Other solubilities: soluble in dmf, methanol
Formula Weight 295.29
Percent Purity 99%
Chemical Name or Material Entecavir monohydrate

RUO – Research Use Only