Learn More
Mopec Ensemble Portable Vented Trash Can, Left
Description
Construction
- Provides a vented trash receptacle solution next to your unit's sink
- Includes trash can assembly on casters, flex duct, (2) clamps, and duct stub in side panel of grossing station unit
Materials
- 304 stainless steel construction with a polished No. 4 finish
Dimensions
- 15.3L x 15.2D x 44.2H in.
Performance
- AX060-L is designed for left-handed Maestro and Maestro Encore grossing stations
Capacity
- 60 lb.
Compatibility
- Fits all left-handed Mopec Maestro and Maestro Encore grossing stations
NOTE: This option must be ordered and at the time of purchase and installed during manufacturing of the grossing station. It cannot be installed in the field after delivery
NOTE: Your lab must have an additional 24 x 24 in. of space next to your grossing station to accommodate this accessory.
Specifications
Specifications
| Includes | Vented Trash Can |