missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Ensemble Interactives Exploring Entropy Kit

Catalog No. S000887
Change view
Click to view available options
Includes:
Classroom Set
Single Set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S000887 Single Set
S000872 Classroom Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Choking Hazard, Prop 65
Catalog No. S000887 Supplier United Scientific™ Supplier No. EIENTI01
Only null left
Choking Hazard, Prop 65

Ensemble Interactives Exploring Entropy Kits are a hands-on demo for understanding entropy.

Students are introduced to several fundamental chemistry and physics concepts, like shake, spin, and rattle containers of different particles to investigate relationships between them. This activity teaches the rigorous definition of entropy by grounding it in direct observations related to everyday experiences regarding spontaneity and disorder. By shaking this model of atoms or other microscopic particles, students observe a surprising and highly memorable disorder-to-order transition, which is easily explained through the concepts learned about entropy. The single set includes one container and particles with sides of: 14 and 20. The classroom set includes seven containers and particles with sides of: 14, 20, 12, 10, 8, 6, and 4. The variety of different sized particles allow for greater variations in results and make the group resultsdistinct from each other. Includes activity guide.

  • Creates opportunities to discuss or introduce concepts of entropy
  • Can be as short as 5-minute instructor's demo or as long as full lab period for many students
  • Learn by direct observation, not analogy
  • Includes activity guide
  • Peer reviewed and patent pending. Featured in the September 2023 issue of 'The Physics Teacher' journal, Volume 61, Issue 6.
  • Avalible in single or class sets

Specifications

Grade 7 to Undergraduate
Includes Single Set
Quantity 7 Containers
Class Size 1 Lab Group
Product Type Entropy Kit