missing translation for 'onlineSavingsMsg'
Learn More

CEP ENPAC™ Spillpal Drum Pallets

Provides incidental spill control during temporary storage of drums, machine parts, tanks and generators

Supplier:  CEP 5755 YE

 View more versions of this product

Catalog No. 19048802


Only null left

Description

Description

  • Flexible four inch foam sidewalls spring back to form, allowing easy drive-in and drive-out with a hand truck or forklift
  • Designed for 37.8L (10 gal.) drums
  • Able to be folded and stored
  • Reusable

Specifications

Chemical Identifiers

Yellow
Foam sidewalls
10.16 cm
Used to store hazardous material
4 in.

Specifications

SPCC, EPA, NPDES
20 gal.
2-drum Spill Pallet
2 Drums
2 x 4 x 4 in.
0.6 m
1.21 m
10.16 cm
1.36 kg
Yellow
Polyethylene
Used to store hazardous material
Foam sidewalls
2 ft.
4 ft.
4 in.
3 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions