missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP ENPAC™ Spillpal Drum Pallets
Provides incidental spill control during temporary storage of drums, machine parts, tanks and generators
Supplier: CEP 5750 YE
Description
- Flexible four inch foam sidewalls spring back to form, allowing easy drive-in and drive-out with a hand truck or forklift
- Designed for 37.8L (10 gal.) drums
- Able to be folded and stored
- Reusable
Chemical Identifiers
| Yellow | |
| Foam sidewalls | |
| 10.16 cm |
| Used to store hazardous material | |
| 4 in. |
Specifications
| SPCC, EPA, NPDES | |
| 10 gal. | |
| 1-drum Spill Pallet | |
| 1 Drum | |
| 2 x 2 x 4 in. | |
| 0.6 m | |
| 0.6 m | |
| 10.16 cm | |
| 0.9 kg |
| Yellow | |
| Polyethylene | |
| Used to store hazardous material | |
| Foam sidewalls | |
| 2 ft. | |
| 2 ft. | |
| 4 in. | |
| 2 lb. |