missing translation for 'onlineSavingsMsg'
Learn More

CEP Enpac™ Poly-Dolly™

Catalog No. 19087809
Change view
Click to view available options
Height (English):
27 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English)
19087809 27 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19087809 Supplier CEP Supplier No. 5300 YE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

U-shaped barrel platform and double-looped strap for smooth, secure loading; kickplate and rolled-grip handles for easy tipping; and large 10 in. dia. (25cm) wheels for trouble-free transport.

Versatile all-in-one dolly, dispensing station and containment unit

Transports 55 gal. (208L) drums with ease.

  • Constructed of tough polyethylene, dolly readily rolls through 36 in. (91cm) doorways and can be handled by a forklift
  • Serves as a secondary containment unit, making mobile dispensing of hazardous liquids safe and efficient
  • Conveniently tilts to dispense liquids; drum contents self-dispense when platform is tilted at 1.5°
  • Dispensing well accommodates 5 gal. (19L) pail; sump has capacity of 80 gal. (303L) with convenient plug to drain contents

Order Info

Exceeds U. S. EPA 40CFR264.175.

Compliance

Exceeds U.S. EPA 40CFR264.175

Specifications

Type Poly-Dolly
Holds 1 Drum (55 gal.)
Load Bearing Capacity (English) 600 lb.
Load Bearing Capacity (Metric) 272 kg
Material Polyethylene
Dimensions (L x W x H) 72 x 32 x 27 in. (183 x 81.28 x 68.58 cm)
For Use With (Application) Mobile dispensing of hazardous liquids
Height (English) 27 in.
Height (Metric) 68.58 cm
Includes U-shaped barrel platform and double looped strap, kickplate and rolled grip handles for easy tipping and large 10 in. dia. wheels
Length (English) 72 in.
Length (Metric) 183 cm
Width (English) 32 in.
Width (Metric) 81.28 cm
Show More Show Less