Learn More
Enoxacin
A quinolone antibacteral agent which acts on DNA gyrase | CAS: 74011-58-8 | C15H17FN4O3 | 320.324 g/mol
Supplier: Thermo Scientific Chemicals J6191206
| Quantity | 5 g |
|---|
Description
Enoxacin is an antibacterial agent used in the treatment of gastro enteritis including infectious diarrhea, respiratory tract infections, gonorrhea and urinary tract infections. It is a bactericidal and its mode of action depends on blocking of bacterial DNA replication by binding itself to an enzyme called DNA gyrase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEnoxacin is an antibacterial agent used in the treatment of gastro enteritis including infectious diarrhea, respiratory tract infections, gonorrhea and urinary tract infections. It is a bactericidal and its mode of action depends on blocking of bacterial DNA replication by binding itself to an enzyme called DNA gyrase.
Solubility
50 mg/ml in 1 M NaOHSlightly soluble in sodium hydroxide, dimethyl sulfoxide, chloroform and methanol. Insoluble in water.
Notes
Store in cool place. Keep the container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
| 74011-58-8 | |
| 320.324 | |
| IDYZIJYBMGIQMJ-UHFFFAOYSA-N | |
| 3229 | |
| 1-ethyl-6-fluoro-4-oxo-7-piperazin-1-yl-1,8-naphthyridine-3-carboxylic acid |
| C15H17FN4O3 | |
| MFCD00133308 | |
| Penetrex™ The Clorox Company; Bactidan | |
| CHEBI:157175 | |
| CCN1C=C(C(=O)C2=CC(=C(N=C21)N3CCNCC3)F)C(=O)O |
Specifications
| Enoxacin | |
| Crystalline Powder | |
| 220°C to 224°C | |
| 5 g | |
| MFCD00133308 | |
| Penetrex™ The Clorox Company; Bactidan | |
| IDYZIJYBMGIQMJ-UHFFFAOYSA-N | |
| 1-ethyl-6-fluoro-4-oxo-7-piperazin-1-yl-1,8-naphthyridine-3-carboxylic acid | |
| 3229 | |
| 320.32 |
| Enoxacin | |
| 74011-58-8 | |
| White to Yellow | |
| C15H17FN4O3 | |
| 14,3587 | |
| 50mg/ml in 1 M NaOHSlightly soluble in sodium hydroxide,dimethyl sulfoxide,chloroform and methanol; Insoluble in water. | |
| CCN1C=C(C(=O)C2=CC(=C(N=C21)N3CCNCC3)F)C(=O)O | |
| 320.324 | |
| CHEBI:157175 |
Safety and Handling
RTECSNumber : QN2800000
TSCA : No
Recommended Storage : Keep cold