Learn More
Eisco™ Enhanced Optical Bench Set
Catalog No. S13478
Change view
Click to view available options
Enhanced optical bench set
Catalog No.
S13478 Enhanced optical bench set
1 options
Catalog No. S13478 Supplier Eisco™ Supplier No. PH0649
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
1m bench, 8 riders with tightening screw, 1 lamp housed with halogen bulb 12 Volt, 24 Watt, 6 Lens Holders for 50mm dia. lenses, +10,+20,+50 Biconvex lens, -20 Biconcave lens, Set of diaphragms, Plain mirror 100 x 100mm, Translucent screen 100 x 100mm, Transparent screen 100 x 100mm, white object screen 100 x 100mm
For performing experiments in optics.
  • Bench graduated on one side
  • Includes halogen lamp
Specifications
Product Type Enhanced Optical Bench Set
For Use With (Application) Performing experiments in optics
Includes 1m bench, 8 riders with tightening screw, 1 lamp housed with halogen bulb 12 Volt, 24 Watt, 6 Lens Holders for 50mm dia. lenses, +10,+20,+50 Biconvex lens, -20 Biconcave lens, Set of diaphragms, Plain mirror 100 x 100mm, Translucent screen 100 x 100mm, Tr