missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Engino™ STEM Solar Power Complete Set

Catalog No. S03052
Change view
Click to view available options
Product Type:
STEM Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S03052 STEM Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

Choking Hazard

Catalog No. S03052 Supplier United Scientific™ Supplier No. UNUNE951

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Choking Hazard

Innovative product line to introduce and master STEM Concepts.

Climate change and global warming urge scientists to take action and save our planet by reducing carbon emissions. The sun can provide free energy for all of our needs, however, we need to convert it to a useful form. This set includes one 3V solar panel that powers an electrical motor, along with digital instructions for 16 solar powered models such as a chopper car, a fan, a plane, a robot and a winch crane. With easy to-follow digital building instructions and detailed explanations of the different scientific principles applied, along with innovative experimental activities, students learn by doing all about Solar Energy while acquiring 21st century skills.

Suggested for students working in groups of 2 or 3.

Technical Specifications:

  • 142 Plastic parts
  • 627 Connecting points
  • 1 DC motor with option to operate in-house with batteries
  • 1 3V Solar panel with cable
  • Digital instructions for 16 models
  • Download the easy-to-follow instructions, theory and quizzes
  • Online step by step building guide
  • Encourages creativity and critical thinking
  • Versatile with the ability to build different models

Specifications

Product Type STEM Kit
Height (English) 11.5 in.
Height (Metric) 29 cm
Length (English) 11.5 in.
Length (Metric) 29 cm