Learn More
United Scientific™ Engino™ STEM and Robotics PRO Set
Description
The STEM and Robotics ERP Pro education set is specially designed for late Primary and Secondary school students of ages 9-12+. It combines the core subjects of STEM together with Robotics and includes high-level projects that lead to a deeper understanding of the underlying theory while also developing 21st century skills. The set comes in a convenient plastic storage tub that contains a large number of ENGINO™ structural and technical parts, together with advanced robotic devices such as ERP Pro controller, DC motors, LEDs, touch sensor and IR sensors. The set allows the construction of more than 30 STEM and Robotic models.
Specifications:- Plastic tub (43 x 31 x 15 cm) with internal dividers
- 369 Plastic parts
- 1678 Connecting points
- Battery compartment for 6 x AA batteries
- Pro controller with 7 input-output ports
- Connectivity via USB and Bluetooth
- Onboard membrane buttons for manual programming
- 3 DC motors
- 5 LEDs
- 2 IR sensors and 1 Touch sensor
- Pedestrian crossing cardboard
- Download the easy-to-follow instructions, theory and quizzes
- Includes sample programs
- Online step by step building guide
- Encourages creativity and critical thinking
- Versatile with the ability to build different models
Specifications
Specifications
| Product Type | STEM Kit |
| Grade Level | High School |