Learn More
United Scientific™ Engino™ STEM and Robotics Mini Set
Innovative product line to introduce and master STEM Concepts.
Supplier:  United Scientific™ UNE201
Catalog No. S03055
Only null left
Alert:
Choking Hazard
Description
Description
The STEM and Robotics ERP Mini education set is specially designed for Early Primary school students of ages 7-9. It combines the core subjects of STEM together with Robotics. The set comes in a convenient plastic storage tub that contains a large number of Engino - Qboidz plastic parts along with robotic devices such as controller Mini, DC motor, LED, touch sensor, IR sensor. The extra ENGINO™ technical parts allow the construction of 40 complex models that come with theory and experimental activities to cover not only engineering design but also Coding, Mechanics and Science. There are two generations of robotic models doable with this set. Simpler models using the Qboidz parts which aim to introduce coding to the younger students, and complex ones with the classic ENGINO parts for more advanced models.
Suggested for students working in groups of 2 or 3.
Specifications:
  • Plastic tub (43 x 31 x 15 cm) with internal dividers
  • 312 Plastic parts
  • 1596 Connecting points
  • Battery compartment for 3 x AAA batteries
  • Mini controller with 4 input-output ports
  • Connectivity via USB and Bluetooth
  • Onboard membrane buttons for manual programming
  • 2 IR sensors and 1 Touch sensor
  • 2 DC motors
  • 1 LED red
  • Download the easy-to-follow instructions, theory and quizzes
  • Includes sample programs
  • Online step by step building guide
  • Encourages creativity and critical thinking
  • Versatile with the ability to build different models
Specifications
Specifications
STEM Kit
Product Suggestions
Product Suggestions