Learn More
United Scientific™ Engino™ STEM Robotics ERP Mini Set
Engino Robotics Platform (ERP) is specially designed for younger students and considers the most modern pedagogical principles of ICT and STEM learning.
Supplier:  United Scientific™ STEM60
Numéro de catalogue. S000840
Il en reste null
Découvrez plus d'offres spéciales
Avertissement:
Choking Hazard
Description
Description
The set consists of all the basic parts for studying robotics! The booklet provides detailed explanations of the different scientific principles applied and incorporates innovative activities for hands-on learning, along with a detailed user manual with programming examples to get you started in Prerecorded programs: The ERP controller contains pre-recorded programs in its memory, designed to control 4 specific models of the set! Build the model you like and set the controller in pre-recorded mode by holding the desired button pressed when turning on the device. The buttons and programs are described inside the User Manual booklet. Manual programming: the most innovative characteristic of the ERP is the onboard buttons, which allow manual programming without the need to connect to a PC. Programs recorded can then be imported to the GUI software and edited either via text language or graphical. Programming software: one of the key elements of any robotics system is programming software. Suggested for a group of 2 to 3 students.
  • ARM Cortex-M4 32-bit processor, 64 MHZ. 512 kB flash/64 kB RAM
  • USB full speed port (12 Mbit/s)
  • Onboard membrane buttons for manual programming
  • 4 input - output ports
  • Connect up to 2 sensors digital or analogue and up to 2 motors (servo or analogue)
  • Power source: 3 x AAA batteries (not included).
  • Includes a manual with instructions, theory and quizzes
  • Online step by step building guide
  • Encourages creativity and critical thinking
  • Versatile with the ability to build different models
Spécifications
Spécifications
STEM Kit
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS