missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Engino™ STEM Paleontology

Catalog No. S000843
Change view
Click to view available options
Grade:
5+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S000843 5+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Choking Hazard
Catalog No. S000843 Supplier United Scientific™ Supplier No. STEM09
Choking Hazard

Learn all about the dinosaurs and how they lived in the prehistoric era on Earth with the Engino STEM Paleontology kit.

Build 5 motorized dinosaur models such as a brontosaurus, a stegosaurus, a triceratops, a pterodactyl and an ankylosaurus and bring the to motion. You can find easy to follow building instructions for all models either online or in the booklet included. The booklet provides detailed explanations of the different scientific principles applied and incorporates innovative experimental activities for hands on learning. A Quiz section is also available to challenge your newly acquired knowledge! Suggested for a group of 2 to 3 students

  • Includes a manual with instructions, theory and quizzes
  • Online step by step building guide
  • Encourages creativity and critical thinking
  • Versatile with the ability to build different models

Specifications

Grade 5+
Class Size 1 Lab Group
Product Type Paleontology Kit