Learn More
United Scientific™ Engino™ STEM Mechanics: Wheels, Axles, and Inclined Planes
Description
Find out how an inclined plane can be used for lifting heavy objects and how another form of inclined plane, the wedge, is used in everyday applications. Build 14 working models such as a launching platform, a door with knob, a well, an airport staircase, an experimental ramp and a splitting wedge. You can find easy-to-follow building instructions for all models either online or in the booklet included. The booklet provides detailed explanations of the different scientific principles applied and incorporates innovative experimental activities for hands-on learning. A Quiz section is also available to challenge your newly acquired knowledge! Suggested for a group of 2 to 3 students.
- Includes a manual with instructions, theory and quizzes
- Online step by step building guide
- Encourages creativity and critical thinking
- Versatile with the ability to build different models
Specifications
Specifications
| Grade | 5+ |
| Class Size | 1 Lab Group |
| Product Type | Simple Machine Kit |