Learn More
United Scientific™ Engino™ STEM Mechanics: Pulley Drives
Catalog No. S03069
Change view
Click to view available options
Type: Pulley Kit
Catalog No.
S03069 Type: Pulley Kit
1 options
Choking Hazard
Catalog No. S03069 Supplier United Scientific™ Supplier No. STEM03
Choking Hazard
Innovative product line to introduce and master STEM Concepts.
Learn how pulley drives can be used to transfer force with reduced friction and how they can increase force or speed at amazing levels. Pulleys have been used for thousands of years and are essential parts of complex machines in modern times, solving many technological problems. Build 8 working models such as a material lift, a stationary bike, a crane bridge, a blender, a construction crane and a windmill. You can find easy-to-follow building instructions for all models either online or in the booklet included. The booklet provides detailed explanations of the different scientific principles applied and incorporates innovative experimental activities for hands-on learning. A Quiz section is also available to challenge your newly acquired knowledge.
  • Includes a manual with instructions, theory and quizzes
  • Online step by step building guide
  • Encourages creativity and critical thinking
  • Versatile with the ability to build different models
Specifications
Product Type Pulley Kit