missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Engino™ STEM Mechanics Master Set

Catalog No. S000833
Change view
Click to view available options
Grade:
5+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S000833 5+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Choking Hazard
Catalog No. S000833 Supplier United Scientific™ Supplier No. UNUNE971

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Choking Hazard

The STEM Mechanics Master set is the ultimate combination of the main subjects of Simple Machines and Physics.

The STEM Mechanics Master set includes Levels, Pulleys, Gears, Linkages, Newton's Laws, Energy conversion, Solar Power and Structures. The patented geometry of ENGINO™ components allows connectivity to all directions of the 3D space, enabling users to easily create technical models. This unique set enhances creativity and imagination while also teaching a variety of STEM principles. A geared 3V motor with a large solar panel is included for animating the ENGINO™ builds, converting solar energy to electricity. The set comes with a library of high-precision gears, pulleys and axles for changing speed, force and direction of motion. From this set, besides open projects, students can find digital instructions for 100 working models and amazing animated contraptions such as cars, cranes, bridges! Suggested for a group of 2 to 3 students

  • Plastic tub (43 x 31 x 15 cm)
  • 283 Plastic parts
  • 1037 Connecting points
  • Digital instructions for 100 models
  • Internal divider
  • 1 DC motor
  • Gearing system
  • Pulleys system
  • 1 x 3 V Solar panel with cable
  • Download the free app to view and manipulate 3D models
  • Encourages creativity and critical thinking
  • Download the easy-to-follow instructions and activity guide
  • Versatile with the ability to build different models

Specifications

Grade 5+
Class Size 1 Lab Group
Product Type Simple Machine Kit