Learn More
United Scientific™ Engino™ STEM MECHANICS: Levers, Linkages and Structures
Catalog No. S03060
Change view
Click to view available options
Type: Lever Model
Catalog No.
S03060 Type: Lever Model
1 options
Choking Hazard
Catalog No. S03060 Supplier United Scientific™ Supplier No. STEM901
Choking Hazard
Innovative product line to introduce and master STEM Concepts.
Learn how Levers are used to increase a force for lifting heavy objects and how you can create models with complex motion by connecting many levers together. Discover all the types of forces acting on structures and find out ways to reduce their effects. Build 16 working models such as a seesaw, a movable weight scale, a wheelbarrow, a parking gate, a toy with moving figures, a pantograph, a cube and a double deck bridge. You can find easy-to-follow building instructions for all models either online or in the booklet included. The booklet provides detailed explanations of the different scientific principles applied and incorporates innovative experimental activities for hands-on learning. A Quiz section is also available to challenge your newly acquired knowledge.
  • Includes a manual with instructions, theory and quizzes
  • Online step by step building guide
  • Encourages creativity and critical thinking
  • Versatile with the ability to build different models
Specifications
Product Type Lever Model