Learn More
United Scientific™ Engino™ STEM Mechanics: Gears and Worm Drives
Catalog No. S03067
Change view
Click to view available options
Type: Kit, Handheld Centripetal Force Kit
Catalog No.
S03067 Type: Kit, Handheld Centripetal Force Kit
1 options
Choking Hazard
Catalog No. S03067 Supplier United Scientific™ Supplier No. STEM05
Choking Hazard
Innovative product line to introduce and master STEM Concepts.
Learn how gears can easily reduce or increase speed, change force or transfer motion from one position to another. Discover how worm drives are used to greatly reduce rotational speed and how screws can convert rotational motion to linear, while greatly increasing force. Build 12 working models such as an experimental crane, a gearbox, a carousel, a helicopter, a screw press and a crane with a rotating arm. You can find easy-to-follow building instructions for all models either online or in the booklet included. The booklet provides detailed explanations of the different scientific principles applied and incorporates innovative experimental activities for hands-on learning. A Quiz section is also available to challenge your newly acquired knowledge.
  • Includes a manual with instructions, theory and quizzes
  • Online step by step building guide
  • Encourages creativity and critical thinking
  • Versatile with the ability to build different models
Specifications
Product Type Kit, Handheld Centripetal Force Kit