missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Engino™ STEM How Cars Work? Technology of Machines

Catalog No. S000837
Change view
Click to view available options
Grade:
5+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S000837 5+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Choking Hazard
Catalog No. S000837 Supplier United Scientific™ Supplier No. STEM23

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Choking Hazard

Learn all about cars with the Engino STEM How Cars Work Kit.

Cars or automobiles, are probably the most common way for transporting humans and goods. They may vary in size and design, but all are defined as motor vehicles that run primarily on roads and have wheels. Cars came into global use during the 20 century and our developed economies depend on them. Who was the inventor of the car? What are the basic parts of a car and how do they work? How crucial is safety and comfort for passengers? Which physical laws car engineers need to know? Suggested for a group of 2 to 3 students.

  • Includes a manual with instructions, theory and quizzes
  • Online step by step building guide
  • Encourages creativity and critical thinking
  • Versatile with the ability to build different models

Specifications

Grade 5+
Class Size 1 Lab Group
Product Type Activity Kit