missing translation for 'onlineSavingsMsg'
Learn More
Learn More
United Scientific™ Engino™ STEM How Aircrafts Work? Technology of Machines
Description
Travelling with an aircraft for holidays or for business is a very common activity. Airplanes are made to combine fast transportation and safety. The development of aviation is considered as one of the highlights in human history, since it has totally changed the way we travel around the Earth. Are you interested to become a pilot? How do pilots control a plane and do acrobatic maneuvers? Which forces affect a flight? How do helicopters fly? Suggested for a group of 2 to 3 students.
- Includes a manual with instructions, theory and quizzes
- Online step by step building guide
- Encourages creativity and critical thinking
- Versatile with the ability to build different models
Specifications
Specifications
| Grade | 5+ |
| Class Size | 1 Lab Group |
| Product Type | Activity Kit |