missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Engino™ STEM Astronomy: Travelling to Space

Catalog No. S03064
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S03064 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

Choking Hazard

Catalog No. S03064 Supplier United Scientific™ Supplier No. STEM08

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Choking Hazard

Innovative product line to introduce and master STEM Concepts.

Learn how rockets are launched into space and how spacecrafts explore the solar system. Learn about the mission to the Moon and how astronauts live in the International Space Station (ISS). Discover the science of space exploration and the hazardous threats of space environment. Build 5 models such as a rocket, a landing pot, scale, a space robot and a space shuttle. You can find easy-to-follow building instructions for all models either online or in the booklet included. The booklet provides detailed explanations of the different scientific principles applied and incorporates innovative experimental activities for hands-on learning. A Quiz section is also available to challenge your newly acquired knowledge.

  • Includes a manual with instructions, theory and quizzes
  • Online step by step building guide
  • Encourages creativity and critical thinking
  • Versatile with the ability to build different models

Specifications

Grade 6 to 12
Format Booklet
Product Type STEM Building Kit
Languages English
Subject Earth Science