Learn More
United Scientific™ Engino™ Makerspace DIY: Hydraulic Robot Arm
The Engino Makerspace DIY Line uses advanced mathematical design with simple to use laser cut wooden parts to create a fun and easy to use design experience.
Supplier: United Scientific™ UNEMS04
Alert:
Choking Hazard
Description
Accompanying the series is a detailed theory book. This resource extends beyond simple assembly instructions to offer a deep dive into the model's technical and scientific principles. It's packed with enjoyable activities that align with the series themes, fostering key 21st century skills. Through this comprehensive approach, ENGINO™ aims to deliver an educational experience that is both enriching and transformative, equipping learners with the tools they need to succeed in a rapidly evolving world. The laser-cut wooden surfaces are connected in stacked formation to create a solid working hydraulic arm with pivoting joints. The hydraulic design is easy to connect with the included tubes and 4 syringes. The geared system allows the arm to be manually rotated to 180°. The finished device is 42 cm long and 22 cm tall. Suggested for a group of 2 to 3 students
- Includes a manual with instructions and background
- Online step by step building guide
- Includes a paint set
- The final product is a real working model
Specifications
| STEM Kit |