missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Engino™ Greenhouse Set

Catalog No. S000832
Change view
Click to view available options
Grade:
5+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S000832 5+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Choking Hazard
Catalog No. S000832 Supplier United Scientific™ Supplier No. UNUNE941

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Choking Hazard

With the Engino Greenhouse Kit, students can construct their very own large greenhouse.

This kit measures 37 x 35 x 25 cm and kids can learn about key principles of biology such as photosynthesis through hands-on experimentation. The greenhouse is designed with opening roof flaps that use a gearing system, allowing students to explore a variety of subjects including science, structures, mechanics, and agronomy. Students can manually control the conditions inside the greenhouse and monitor how their plants respond to changes in temperature, moisture, and light levels. What's more, this kit is highly customizable and can be extended to become fully autonomous by adding DIY controllers and sensors. Whether you're a teacher looking for a fun and engaging way to teach your students about sustainability or a parent looking for an educational adventure for your child, this kit is perfect for you! Suggested for a group of 2 to 3 students

  • Download the free app to view and manipulate 3D models
  • Encourages creativity and critical thinking
  • Download the easy-to-follow instructions and activity guide

Specifications

Grade 5+
Class Size 1 Lab Group
Product Type Botany Classroom Kits