Learn More
United Scientific™ Engino™ GINOBOT Basic Edition
Engino™ GINOBOT Basic Edition is an innovative product line to introduce and master STEM Concepts.
Supplier:  United Scientific™ UNE491
Catalog No. S03053
Only null left
Alert:
Choking Hazard
Description
Description
GinoBot™ is a highly programmable robot ready to be used straight out of the box. Developed by a high-level research team of engineers and academics, GinoBot™ is a unique tool for teaching STEM disciplines, computational thinking and digital literacy with fun and hands-on experiential activities. The body of GinoBot™ has numerous built-in ENGINO™ connecting geometries so that students can expand their robot with structural and mechanical parts, making more advanced models. Teachers can follow STEM interdisciplinary activities and apply inquiry-based learning methodologies to engage and inspire their students. The vehicle's powerful processor connects with USB, Bluetooth and Wi-Fi and can be programmed with KEIRO, but also Python and C++ once a Micro:bit or Arduino are installed. The 4-wheeled robot can have its tires converted to a cat-truck and is surrounded by three proximity sensors. It has two floor color sensors and two encoder motors for precise motion. Suggested for students working in groups of 3 or 4.
Technical Specifications:
  • 16 Plastic parts
  • 87 Connecting points
  • Download the easy-to-follow instructions, theory and quizzes
  • Includes sample programs
  • Online step by step building guide
  • Encourages creativity and critical thinking
  • Versatile with the ability to build different models
Specifications
Specifications
STEM Kit
Product Suggestions
Product Suggestions