missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENG Scientific Systain™ Methanol
Systain™ Methanol is for use with Sysmex™ hematology slide stainer.
$783.53 - $783.53
Specifications
| Storage Requirements | Room Temperature |
|---|---|
| For Use With (Application) | For in-vitro diagnostic use only, use as fixative |
| For Use With (Equipment) | For Sysmex hematology slide stainer |
Description
- Use as fixative prior to staining specimen material
- In the event of cold weather, allow solutions to return to room temperature
Specifications
| Room Temperature | |
| For in-vitro diagnostic use only, use as fixative | |
| For Sysmex hematology slide stainer |
For in vitro diagnostic use only.