Learn More
ENG Scientific Reticulocyte Stain DFS Item
Use for finger sticks or anticoagulated blood. Mix equal volumes of Reticulocyte Stain (New Methylene Blue (NMB)) with blood. No counterstaining necessary.
$718.20 - $1080.33
Specifications
For Use With (Application) For in-vitro diagnostic use only, may be used for finger sticks or anticoagulated Blood
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Type Price Quantity  
Catalog Number Mfr. No. Quantity Type Price Quantity  
ES5100 DFS Item
View Documents
ENG Scientific
5100
2 x 100 mL NMB
Pack of 2 Each for $1,080.33
Only null left
ES5101 DFS Item
View Documents
ENG Scientific
5101
1 x 100 mL NMB
Each for $718.20
Only null left
Description
Description
  • The proportion of blood to stain should be increased for low hemoglobins.
  • May be used for finger sticks or anticoagulated blood.
Specifications
Specifications
For in-vitro diagnostic use only, may be used for finger sticks or anticoagulated Blood
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
For in vitro diagnostic use only.