missing translation for 'onlineSavingsMsg'
Learn More

ENG Scientific Lactophenol Cotton Blue Stain, Certified

Catalog No. ES6730
Encompass
Change view
Click to view available options
Quantity:
2 x 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES6730 2 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ES6730 Supplier ENG Scientific Supplier No. ES6730

For staining fungi using Lactophenol Cotton Blue Stain.

  • Lactic acid acts as a preservative for fungi, killed by phenol and stained by cotton blue
  • Fungal elements are stained a deep blue; background is pale blue
  • Lactophenol Cotton Blue Stain
  • Includes Phenol, lactic acid, glycerol, aniline blue
  • For use with staining fungi, in-vitro diagnostic use/li>

Specifications

Quantity 2 x 100 mL
Includes Phenol, lactic acid, glycerol, aniline blue
Format Bottle
For Use With (Application) Staining fungi, in-vitro diagnostic use
Stain Type Lactophenol Cotton Blue
Product Type Lactophenol Cotton Blue Stain

For in vitro diagnostic use only.