missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ RevoLite™ 40 cal Arc Flash Kit with Lift Front Hood, No Gloves

Catalog No. 170011941
Change view
Click to view available options
Size:
4X-Large
5X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
170011941 5X-Large
170011940 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 170011941 Supplier National Safety Apparel™ Supplier No. KIT4SCLT40NGLF5X

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAT 4 Arc Flash Kit includes 40 Cal RevoLite™ short coat and bib overall, lift front hood with PureView™ faceshield and hard hat, safety glasses, hood bag, and gear bag.

  • 50% Lighter than our standard 40 cal kits
  • FR hook and loop front closure
  • Stand up collar for extra coverage
  • Rib knit cuffs for comfortable, secure fit
  • Garments embroidered with arc rating
  • Quick disconnect and adjustable shoulder straps on bib overalls
  • Expandable leg opening to fit over boots
  • Gloves are not included

Specifications

Gender Unisex
Suspender None
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Seam Type Stitched
Material RevoLite™
Closure Type FR Hook and Loop Front Closure, Buckle
Size 5X-Large
Arc Flash Rating 40 cal/cm²
Certifications/Compliance NFPA 70E, ASTM F1506, CSA Z462, OSHA 1910.269
Color Olive Green
Product Type Arc Flash Kit
Show More Show Less