missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ RevoLite™ 40 cal Arc Flash Kit with Lift Front Hood, No Gloves

Catalog No. 170011940
Change view
Click to view available options
Size:
4X-Large
5X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
170011940 4X-Large
170011941 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 170011940 Supplier National Safety Apparel™ Supplier No. KIT4SCLT40NGLF4X

CAT 4 Arc Flash Kit includes 40 Cal RevoLite™ short coat and bib overall, lift front hood with PureView™ faceshield and hard hat, safety glasses, hood bag, and gear bag.

  • 50% Lighter than our standard 40 cal kits
  • FR hook and loop front closure
  • Stand up collar for extra coverage
  • Rib knit cuffs for comfortable, secure fit
  • Garments embroidered with arc rating
  • Quick disconnect and adjustable shoulder straps on bib overalls
  • Expandable leg opening to fit over boots
  • Gloves are not included

Specifications

Gender Unisex
Color Olive Green
Suspender None
Arc Flash Rating 40 cal/cm²
Certifications/Compliance NFPA 70E, ASTM F1506, CSA Z462, OSHA 1910.269
Product Type Arc Flash Kit
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Closure Type FR Hook and Loop Front Closure, Buckle
Material RevoLite™
Seam Type Stitched
Size 4X-Large
Show More Show Less