missing translation for 'onlineSavingsMsg'
Learn More

Supertek™ Energy Conversion Kit
GREENER_CHOICE

Catalog No. S160129
Encompass
Change view
Click to view available options
Grade:
9 to Undergraduate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S160129 9 to Undergraduate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S160129 Supplier Supertek™ Supplier No. PH20448

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This easy to use Supertek™ Energy Conversion Kit is used to demonstrate the energy conversion between different forms of energy (solar, mechanical, electrical etc.).

  • Easy to use.
  • ON/OFF switch.
  • Solar cell.
  • AA battery holder with batteries.
  • Hand crank.
  • Mounted on a solid plastic board.
  • 4 mm sockets are provided for the out put, which includes a buzzer, led, electromagnet, motor with fan, and a resistor

Specifications

Grade 9 to Undergraduate
Format Kit
Languages English
Subject Physical Science - Energy Conversion