missing translation for 'onlineSavingsMsg'
Learn More

Superior Glove™ Endura™ Arc-Flash Grain Goatskin Leather Drivers Gloves

Catalog No. 17489672
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17489672 Large
17489670 Small
17489671 Medium
17489673 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17489672 Supplier Superior Glove™ Supplier No. 378GKTFGL
Only null left

High tensile strength grain goatskin leather provides abrasion resistance, with a Aramid Fiber liner for ANSI A4 cut protection.

  • Grain goatskin leather has a high tensile strength for abrasion resistance
  • Aramid Fiber provides 360° cut protection
  • Cut and sewn liner ensures a no-bunch custom fit
  • Keystone thumb enhances durability and comfort
  • Arc flash level 3 protection, with an Arc Thermal Performance Value (ATPV) of 36 cal/cm2

Specifications

Arc Flash Level 3
Construction Cut and Sewn
Coating Material Uncoated
Color White
Cut Resistance Rating ANSI A4
Hand Specific
Material Grain Goatskin Leather
Product Type Driver Gloves
Size Large
For Use With (Application) Appliance Manufacturing, Construction, General Maintenance, and Steel Plants
Glove Cuff Style Elasticated
Liner Material Aramid Fiber
Puncture Rating ANSI/ISEA 105-16 Level 3
Series 378GKTFG
Thumb Style Keystone
Show More Show Less